Anti-NPY5R Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB06769-B
EVBIEB06769-BEA 339 EUR
EVBIEB06769-B
Anti-NPY5R Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog