Anti-LRP5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06771
EVBIEB06771EA 335 EUR
EVBIEB06771
Anti-LRP5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRP5 (low density lipoprotein receptor-related protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR