Anti-CFH Goat polyclonal antibody unconjugated
Lieferant: Everest Biotech
EB06848
EVBIEB06848EA 335 EUR
EVBIEB06848
Anti-CFH Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CFH (complement factor H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human