Anti-CFC1 Goat polyclonal antibody unconjugated
Lieferant: Everest Biotech
EB06894
EVBIEB06894EA 339 EUR
EVBIEB06894
Anti-CFC1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CFC1 (cripto, FRL-1, cryptic family 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat