Anti-ADORA2B Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06936
EVBIEB06936EA 339 EUR
EVBIEB06936
Anti-ADORA2B Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADORA2B (adenosine A2b receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR