Anti-ENPP1 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB06948
EVBIEB06948EA 339 EUR
EVBIEB06948
Anti-ENPP1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR