Anti-UCP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06994
EVBIEB06994EA 339 EUR
EVBIEB06994
Anti-UCP1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UCP1 (uncoupling protein 1 (mitochondrial, proton carrier))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR