Anti-FOXP3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07034
EVBIEB07034EA 339 EUR
EVBIEB07034
Anti-FOXP3 Goat Polyclonal Antibody
Antikörper
Western Blot: Not yet tested. At this stage we are dependent on researchers in the field for further characterization of this product. Therefore we cannot recommend an optimal concentration and the antibody is an aspiring product.

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR