Anti-SFN Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB07061-B
EVBIEB07061-BEA 335 EUR
EVBIEB07061-B
Anti-SFN Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: SFN
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig