Anti-ERG Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07081
EVBIEB07081EA 339 EUR
EVBIEB07081
Anti-ERG Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR