Anti-Semaphorin 5A Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB07352
EVBIEB07352EA 339 EUR
EVBIEB07352
Anti-Semaphorin 5A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Semaphorin 5A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen