Anti-ALOX5AP Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB07434-B
EVBIEB07434-BEA 339 EUR
EVBIEB07434-B
Anti-ALOX5AP Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: ALOX5AP
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Dog