Anti-ARSA Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07457
EVBIEB07457EA 339 EUR
EVBIEB07457
Anti-ARSA Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARSA (arylsulfatase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR