Anti-PICK1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07467
EVBIEB07467EA 339 EUR
EVBIEB07467
Anti-PICK1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PICK1 (protein interacting with PRKCA 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR