Anti-TCF3 Goat Polyclonal Antibody (Biotinylated)

Lieferant: Everest Biotech
EB07587-B
EVBIEB07587-BEA 339 EUR
EVBIEB07587-B
Anti-TCF3 Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: TCF3
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR