Anti-IER5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07606
EVBIEB07606EA 335 EUR
EVBIEB07606
Anti-IER5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IER5 (immediate early response 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR