Anti-Fumarase / FH Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07874
EVBIEB07874EA 339 EUR
EVBIEB07874
Anti-Fumarase / FH Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Fumarase / FH
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR