Anti-BHMT Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB07943
EVBIEB07943EA 339 EUR
EVBIEB07943
Anti-BHMT Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR