Anti-PTCH1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07960
EVBIEB07960EA 339 EUR
EVBIEB07960
Anti-PTCH1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR