Anti-Her6 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08009
EVBIEB08009EA 202 EUR
EVBIEB08009
Anti-Her6 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Her6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR