Anti-STS Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08062
EVBIEB08062EA 339 EUR
EVBIEB08062
Anti-STS Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STS (steroid sulfatase (microsomal), isozyme S)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR