Anti-PRRX1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08175
EVBIEB08175EA 339 EUR
EVBIEB08175
Anti-PRRX1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRRX1 (paired related homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR