Anti-CYTH2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08181
EVBIEB08181EA 339 EUR
EVBIEB08181
Anti-CYTH2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYTH2 (cytohesin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR