Anti-GOT1 Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB08213-B
EVBIEB08213-BEA 339 EUR
EVBIEB08213-B
Anti-GOT1 Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: GOT1
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat