Anti-SOX8 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08216
EVBIEB08216EA 194 EUR
EVBIEB08216
Anti-SOX8 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOX8 (SRY (sex determining region Y)-box 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR