Anti-EPB41L4A Goat polyclonal antibody unconjugated
Lieferant: Everest Biotech
EB08238
EVBIEB08238EA 194 EUR
EVBIEB08238
Anti-EPB41L4A Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EPB41L4A (erythrocyte membrane protein band 4.1 like 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse