Anti-APOBEC3D Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB08270
EVBIEB08270EA 194 EUR
EVBIEB08270
Anti-APOBEC3D Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: APOBEC3D (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR