Anti-PAM / PAL Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB08374
EVBIEB08374EA 335 EUR
EVBIEB08374
Anti-PAM / PAL Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAM / PAL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human