Anti-RASSF8 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08395
EVBIEB08395EA 335 EUR
EVBIEB08395
Anti-RASSF8 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RASSF8 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR