Anti-LYPLAL1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08406
EVBIEB08406EA 335 EUR
EVBIEB08406
Anti-LYPLAL1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LYPLAL1 (lysophospholipase-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR