Anti-PTN Goat Polyclonal Antibody (Biotinylated)

Lieferant: Everest Biotech
EB08421-B
EVBIEB08421-BEA 339 EUR
EVBIEB08421-B
Anti-PTN Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: PTN
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR