Anti-AICDA Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08451
EVBIEB08451EA 339 EUR
EVBIEB08451
Anti-AICDA Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AICDA (activation-induced cytidine deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR