Anti-SERPINA12 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08542
EVBIEB08542EA 335 EUR
EVBIEB08542
Anti-SERPINA12 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA12 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR