Anti-ERN1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08559
EVBIEB08559EA 335 EUR
EVBIEB08559
Anti-ERN1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ERN1 (endoplasmic reticulum to nucleus signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR