Anti-EPM2A Goat Polyclonal Antibody (Biotinylated)

Lieferant: Everest Biotech
EB08570-B
EVBIEB08570-BEA 335 EUR
EVBIEB08570-B
Anti-EPM2A Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: EPM2A
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Cow
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR