Anti-AVPR2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08787
EVBIEB08787EA 194 EUR
EVBIEB08787
Anti-AVPR2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AVPR2 (arginine vasopressin receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR