Anti-Mcp10 / Mcp9 Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB08817
EVBIEB08817EA 194 EUR
EVBIEB08817
Anti-Mcp10 / Mcp9 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Mcp10 / Mcp9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse