Anti-CHRNA7 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB08853
EVBIEB08853EA 335 EUR
EVBIEB08853
Anti-CHRNA7 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CHRNA7 (cholinergic receptor, nicotinic, alpha 7 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR