Anti-SHBG Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09042
EVBIEB09042EA 335 EUR
EVBIEB09042
Anti-SHBG Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SHBG (sex hormone-binding globulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR