Anti-PTMA Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09070
EVBIEB09070EA 335 EUR
EVBIEB09070
Anti-PTMA Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTMA (prothymosin, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR