Anti-GCKR Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB09275
EVBIEB09275EA 335 EUR
EVBIEB09275
Anti-GCKR Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GCKR (glucokinase (hexokinase 4) regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR