Anti-DLX5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09284
EVBIEB09284EA 335 EUR
EVBIEB09284
Anti-DLX5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DLX5 (distal-less homeobox 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR