Anti-CLEC4C Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB09343
EVBIEB09343EA 339 EUR
EVBIEB09343
Anti-CLEC4C Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CLEC4C (C-type lectin domain family 4, member C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR