Anti-MTHFR Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09354
EVBIEB09354EA 339 EUR
EVBIEB09354
Anti-MTHFR Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MTHFR (methylenetetrahydrofolate reductase (NAD(P)H))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR