Anti-FMR1 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB09362
EVBIEB09362EA 339 EUR
EVBIEB09362
Anti-FMR1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR