Anti-IL18 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09393
EVBIEB09393EA 335 EUR
EVBIEB09393
Anti-IL18 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL18 (interleukin 18 (interferon-gamma-inducing factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR