Anti-STK39 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09416
EVBIEB09416EA 335 EUR
EVBIEB09416
Anti-STK39 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR