Anti-HCAR1 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB09442
EVBIEB09442EA 335 EUR
EVBIEB09442
Anti-HCAR1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HCAR1 (hydroxycarboxylic acid receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR