Anti-NEBL / nebulette Goat Polyclonal Antibody
Lieferant: Everest Biotech
EB09459
EVBIEB09459EA 194 EUR
EVBIEB09459
Anti-NEBL / nebulette Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEBL / nebulette
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse