Anti-UBE2C Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09517
EVBIEB09517EA 339 EUR
EVBIEB09517
Anti-UBE2C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR